Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Acetylthiophene 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methyl 2-thienyl ketone 1-(2-Thienyl)-1-ethanone

A13062-22 A13062-36 A13062-22 A13062-36
AAA13062-36EA 172.99 USD InStock
AAA13062-36 AAA13062-22 CAAAA13062-36 CAAAA13062-22
2-Acetylthiophene 99%
2-Acetylthiophene


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 10…12 °C
Density: 1.168
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty