Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Chloroethoxy)ethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

A13074-18 A13074-30 A13074-0B A13074-30 A13074-18 A13074-0B
AAA13074-0BEA 534.99 USD InStock
CAAAA13074-18 AAA13074-0B AAA13074-30 CAAAA13074-0B AAA13074-18 CAAAA13074-30
2-(2-Chloroethoxy)ethanol 99%
2-(2-Chloroethoxy)ethanol


Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 185 °C
Density: 1.180
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: "9",
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty