Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercapto-1-methylimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Mercapto-1-methyl-1H-imidazole 2-Mercapto-N-methyl-1H-imidazole 2-Mercapto-N-methylimidazole Thiamazole Methimazole

A13094-22 A13094-14 A13094-14 A13094-22
AAA13094-22EA 209.83 USD InStock
CAAAA13094-14 AAA13094-22 AAA13094-14 CAAAA13094-22
2-Mercapto-1-methylimidazole 98%
2-Mercapto-1-methylimidazole
Possesses antioxidant properties


Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 280 °C (dec.)
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00179321
CAS Number: 60-56-0
EINECS: 200-482-4
UN: 3335
ADR: "9",
Merck Index: 13,06000
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty