Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Arabitol 98%

Supplier: Thermo Scientific Chemicals

A13103-14 A13103-06 A13103-22 A13103-22 A13103-14 A13103-06
AAA13103-14EA 185.99 USD InStock
AAA13103-14 AAAA13103-22 AAA13103-06 CAAAAA13103-22 CAAAA13103-06 CAAAA13103-14
L(-)-Arabitol 98%
L(-)-Arabitol


Formula: C₅H₁₂O₅
Melting Pt: 98…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00064290
CAS Number: 7643-75-6
EINECS: 231-582-6
Order Now
Specification Test Results
Optical rotation -11° (c=5 in 8% borax soln)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty