Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Glucono-1,5-lactone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Gluconic acid-1,5-lactone D-Gluconic acid-δ-lacton D-Glucono-δ-lactone D-Gluconic acid lactone GDL

A13105-30 A13105-30 A13105-36 A13105-36 A13105-0E A13105-0E
AAAA13105-30EA 35.99 USD InStock
CAAAAA13105-30 CAAAA13105-36 AAAA13105-30 AAA13105-36 AAA13105-0E CAAAA13105-0E
D-Glucono-1,5-lactone 99%
D-Glucono-1,5-lactone


Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 154…159 °C (dec.)
Density: 1.61 g/cm³ (–5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty