Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylquinoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13109-14 A13109-22 A13109-14 A13109-06 A13109-06
AAA13109-14EA 308.99 USD InStock
AAA13109-14 CAAAAA13109-22 AAA13109-06 CAAAA13109-14 CAAAA13109-06
3-Methylquinoline 98+%
3-Methylquinoline


Formula: C₁₀H₉N
Boiling Pt: 252…253 °C
Melting Pt: 15 °C
Density: 1.069
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014661
CAS Number: 612-58-8
EINECS: 210-315-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty