Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dimethylchlorosilane

A13113-AK A13113-AD
AAA13113-AKEA 205.99 USD InStock
AAA13113-AK AAA13113-AD
Chlorodimethylsilane 97%
Chlorodimethylsilane


Formula: C₂H₇ClSi
Boiling Pt: 34…36 °C
Melting Pt: –111 °C
Density: 0.869
Flash Pt: -28 °C(-18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000495
CAS Number: 1066-35-9
EINECS: 213-912-0
UN: 2924
ADR: "3",III
REACH: 01-2119486782-25
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty