Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Nitrobenzyl bromide α-Bromo-o-nitrotoluene

A13127-06 A13127-22 A13127-06 A13127-14 A13127-22 A13127-14
AAA13127-06EA 60.09 USD InStock
CAAAA13127-22 CAAAA13127-14 AAA13127-06 AAA13127-14 AAA13127-22 CAAAA13127-06
α-Bromo-2-nitrotoluene 98+%
α-Bromo-2-nitrotoluene


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 45…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty