Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoisophthalic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

A13152-22 A13152-14 A13152-06 A13152-06 A13152-14 A13152-22
AAA13152-06EA 75.29 USD InStock
CAAAA13152-22 CAAAA13152-14 AAA13152-06 AAA13152-14 AAA13152-22 CAAAA13152-06
4-Bromoisophthalic acid 96%
4-Bromoisophthalic acid


Formula: C₈H₅BrO₄
MW: 245.03 g/mol
Melting Pt: 296…302 °C
Storage Temperature: Ambient
MDL Number: MFCD00002404
CAS Number: 6939-93-1
EINECS: 230-078-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty