Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Epoxycyclohexane 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclohexene oxide Epoxycyclohexane

A13185-22 A13185-0E A13185-22 A13185-0E A13185-36 A13185-36
AAA13185-22EA 53.29 USD InStock
CAAAA13185-0E AAA13185-22 CAAAA13185-36 CAAAA13185-22 AAA13185-0E AAA13185-36
1,2-Epoxycyclohexane 98+%
1,2-Epoxycyclohexane


Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129…130 °C
Melting Pt: –40 °C
Density: 0.972
Flash Pt: 24 °C(75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty