Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans,trans-2,4-Hexadienal predominantly trans 95%

Supplier: Thermo Scientific Chemicals
Danger

A13200-14 A13200-06 A13200-22 A13200-06 A13200-14 A13200-22
AAA13200-22EA 173.99 USD InStock
CAAAA13200-22 CAAAA13200-14 AAA13200-22 CAAAA13200-06 AAA13200-14 AAA13200-06
trans,trans-2,4-Hexadienal predominantly trans 95%
trans,trans-2,4-Hexadienal


Formula: C₆H₈O
Boiling Pt: 170…173 °C
Density: 0.871
Flash Pt: 67 °C(152 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007004
CAS Number: 142-83-6
EINECS: 205-564-3
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty