Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(1H-Imidazol-1-yl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(1-Imidazolyl)phenol4-(Imidazol-1-yl)phenol

A13211-14 A13211-03 A13211-03 A13211-06 A13211-14
AAAA13211-03EA 30.29 USD InStock
CAAAA13211-14 AAAA13211-03 AAAA13211-06 AAA13211-14 CAAAAA13211-03
4-(1H-Imidazol-1-yl)phenol 97%
4-(1H-Imidazol-1-yl)phenol


Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00005281
CAS Number: 10041-02-8
EINECS: 233-121-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty