Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Aminopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Pyridylamine Pyridin-3-amine

A13216-18 A13216-18 A13216-0B A13216-30 A13216-30 A13216-0B
AAA13216-0BEA 401.99 USD InStock
CAAAA13216-18 AAA13216-0B AAA13216-30 CAAAA13216-30 AAA13216-18 CAAAA13216-0B
3-Aminopyridine 99%
3-Aminopyridine


Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 250…252 °C
Melting Pt: 58…63 °C
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: "6.1",II
Merck Index: 13,00472
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty