Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl terephthalate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Terephthalic acid dimethyl ester

A13220-36 A13220-0E A13220-36 A13220-0E
AAA13220-36EA 39.05 USD InStock
CAAAA13220-36 CAAAA13220-0E AAA13220-36 AAA13220-0E
Dimethyl terephthalate 99%
Dimethyl terephthalate


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 288 °C
Melting Pt: 140…143 °C
Density: 1.290
Flash Pt: 146 °C(294 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty