Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nonenoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A13228-22 A13228-22 A13228-14 A13228-14
AAA13228-14EA 117.99 USD InStock
AAA13228-14 AAAA13228-22 CAAAAA13228-22 CAAAA13228-14
2-Nonenoic acid 98%
2-Nonenoic acid


Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Boiling Pt: 130…132 °C/2 mm
Density: 0.93
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014020
CAS Number: 3760-11-0
EINECS: 223-171-5
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty