Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetaldehyde hydrate ≥75% in water, tech.

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2,2-Trifluoro-1,1-ethanediolTrifluoroacetaldehyde hydrate

A13234-18 A13234-09 A13234-18 A13234-09
AAA13234-09EA 107.99 USD InStock
AAA13234-09 AAA13234-18 CAAAA13234-09 CAAAA13234-18
Trifluoroacetaldehyde hydrate ≥75% in water, tech.
Trifluoroacetaldehyde hydrate


Formula: C₂H₃F₃O₂
MW: 116.039 g/mol
Boiling Pt: 104…106 °C
Density: 1.44
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00167359
CAS Number: 421-53-4
EINECS: 207-006-4
UN: 1989
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty