Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

A13241-22 A13241-22 A13241-36 A13241-30 A13241-14 A13241-14 A13241-36 A13241-30
AAA13241-14EA 41.49 USD InStock
AAA13241-14 AAA13241-36 AAAA13241-30 AAA13241-22 CAAAA13241-22 CAAAAA13241-30 CAAAA13241-36 CAAAA13241-14
2-Bromopyridine 99%
2-Bromopyridine


Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192…194 °C
Melting Pt: –40.1 °C
Density: 1.625
Flash Pt: 97 °C(207 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
EINECS: 203-641-6
UN: 2929
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty