Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propiolic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Propynoic acid

A13245-14 A13245-14 A13245-22 A13245-06 A13245-06 A13245-22
AAA13245-06EA 46.78 USD InStock
CAAAA13245-22 AAA13245-06 AAA13245-14 CAAAA13245-06 AAA13245-22 CAAAA13245-14
Propiolic acid 98+%
Propiolic acid


Formula: C₃H₂O₂
MW: 70.05 g/mol
Boiling Pt: 102 °C/200 mm
Melting Pt: 18 °C
Density: 1.136
Flash Pt: 58 °C(136 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004360
CAS Number: 471-25-0
EINECS: 207-437-8
UN: 2920
ADR: "8",II
Merck Index: 12,08006
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty