Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A13254-36 A13254-14 A13254-14 A13254-22 A13254-22 A13254-36
AAA13254-14EA 54.69 USD InStock
AAA13254-14 AAA13254-36 CAAAA13254-22 AAA13254-22 CAAAA13254-36 CAAAA13254-14
3-Chlorobenzaldehyde 97%
3-Chlorobenzaldehyde


Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 16…18 °C
Density: 1,239
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty