Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methyl-2-furaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Methylfurfural

A13264-36 A13264-22 A13264-22 A13264-36 A13264-14 A13264-14
AAA13264-36EA 578.92 USD InStock
AAA13264-36 AAA13264-14 AAA13264-22 CAAAA13264-36 CAAAA13264-14 CAAAA13264-22
5-Methyl-2-furaldehyde 98%
5-Methyl-2-furaldehyde


Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 186…188 °C
Density: 1.106
Flash Pt: 72 °C(161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty