Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetaldehyde diethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-1,1-diethoxyethane

A13276-0E A13276-22 A13276-36 A13276-22 A13276-0E A13276-36
AAA13276-36EA 120.99 USD InStock
CAAAA13276-0E CAAAA13276-36 AAA13276-36 AAA13276-0E CAAAA13276-22 AAA13276-22
Bromoacetaldehyde diethyl acetal 97%
Bromoacetaldehyde diethyl acetal


Formula: BrH₂CCH(OCH₂CH₃)₂
MW: 197.07 g/mol
Boiling Pt: 66…67 °C/18 mm
Density: 1.270
Flash Pt: 61 °C(142 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000214
CAS Number: 2032-35-1
EINECS: 217-989-1
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty