Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Picolyl chloride hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-(Chloromethyl)pyridine hydrochloride3-(Chloromethyl)-pyridinium chloride

A13279-06 A13279-06 A13279-14 A13279-14 A13279-22 A13279-22
AAA13279-06EA 30.79 USD InStock
AAA13279-06 CAAAA13279-22 CAAAA13279-14 CAAAA13279-06 AAA13279-14 AAA13279-22
3-Picolyl chloride hydrochloride 97%
3-Picolyl chloride hydrochloride


Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 136…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty