Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloronicotinonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-5-cyanopyridine 2-Chloropyridine-5-carbonitrile

A13291-06 A13291-14 A13291-06 A13291-03 A13291-14 A13291-03
AAA13291-14EA 246.99 USD InStock
AAA13291-14 AAA13291-03 AAA13291-06 CAAAA13291-14 CAAAA13291-03 CAAAA13291-06
6-Chloronicotinonitrile 98%
6-Chloronicotinonitrile


Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Boiling Pt: 105…107 °C/1 mm
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00084941
CAS Number: 33252-28-7
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty