Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethyl phosphonoacetate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Methyl 2-(dimethoxyphosphoryl)acetateDimethyl methoxycarbonylmethanephosphonate

A13301-14 A13301-22 A13301-36 A13301-22 A13301-14 A13301-36
AAA13301-22EA 127 USD InStock
CAAAA13301-14 CAAAA13301-36 CAAAA13301-22 AAA13301-22 AAA13301-36 AAA13301-14
Trimethyl phosphonoacetate 98%
Trimethyl phosphonoacetate


Formula: C₅H₁₁O₅P
MW: 182.11 g/mol
Boiling Pt: 119…120 °C/1 mm
Density: 1.263
Flash Pt: 70 °C(158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008452
CAS Number: 5927-18-4
EINECS: 227-663-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty