Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromothiophene-2-carbaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromo-2-thiophenecarboxaldehyde4-Bromothiophene-2-carboxaldehyde

A13304-14 A13304-14 A13304-06 A13304-22 A13304-22 A13304-06
AAA13304-22EA 435.99 USD InStock
CAAAA13304-22 CAAAA13304-14 CAAAA13304-06 AAA13304-22 AAA13304-06 AAA13304-14
4-Bromothiophene-2-carbaldehyde 96%
4-Bromothiophene-2-carbaldehyde


Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 114…115 °C/11 mm
Melting Pt: 44…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005431
CAS Number: 18791-75-8
EINECS: 242-577-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty