Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: m-Bromobenzonitrile

A13317-18 A13317-30 A13317-18 A13317-09 A13317-30 A13317-09
AAA13317-09EA 77.49 USD InStock
AAA13317-09 CAAAA13317-09 CAAAA13317-18 AAA13317-18 CAAAAA13317-30 AAAA13317-30
3-Bromobenzonitrile 99%
3-Bromobenzonitrile


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 224…226 °C
Melting Pt: 37…41 °C
Density: 1.562
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty