Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Quinuclidinone hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

A13320-06 A13320-14 A13320-22 A13320-22 A13320-06 A13320-14
AAA13320-22EA 226.99 USD InStock
AAA13320-22 CAAAA13320-22 CAAAA13320-06 CAAAA13320-14 AAA13320-06 AAA13320-14
3-Quinuclidinone hydrochloride 98+%
3-Quinuclidinone hydrochloride


Formula: C₇H₁₂ClNO
Melting Pt: ca 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty