Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methyl-5-hepten-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-2-hepten-6-one

A13335-22 A13335-14 A13335-14 A13335-22
AAAA13335-14EA 26.49 USD InStock
CAAAAA13335-14 AAAA13335-14 AAA13335-22 CAAAA13335-22
6-Methyl-5-hepten-2-one 98%
6-Methyl-5-hepten-2-one


Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –67 °C
Density: 0.855
Flash Pt: 56 °C(133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008905
CAS Number: 110-93-0
EINECS: 203-816-7
UN: 1224
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty