Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylcinnamic acid predominantly trans 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(p-Tolyl)acrylic acid

A13337-06 A13337-22 A13337-14 A13337-14 A13337-22 A13337-06
AAA13337-06EA 30.69 USD InStock
AAA13337-06 CAAAA13337-06 AAA13337-14 AAA13337-22 CAAAA13337-22 CAAAA13337-14
4-Methylcinnamic acid predominantly trans 99%
4-Methylcinnamic acid


Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002697
CAS Number: 1866-39-3
EINECS: 217-479-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty