Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Tolylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Methylphenyl)metaboric acid 4-Methylbenzeneboronic acid 4-Methylphenylboronic acid Dihydroxy(4-methylphenyl)borane

A13347-06 A13347-14 A13347-14 A13347-06 A13347-03 A13347-03
AAA13347-14EA 447.99 USD InStock
AAA13347-14 AAA13347-03 CAAAA13347-06 AAA13347-06 CAAAA13347-14 CAAAA13347-03
4-Tolylboronic acid 98%
4-Tolylboronic acid


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 246…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00039138
CAS Number: 5720-05-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty