Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrazine-2-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Pyrazinecarboxylic AcidPyrazinecarboxylic acid

A13363-22 A13363-14 A13363-22 A13363-14
AAA13363-14EA 42.99 USD InStock
AAA13363-14 AAA13363-22 CAAAA13363-14 CAAAA13363-22
Pyrazine-2-carboxylic acid 99%
Pyrazine-2-carboxylic acid


Formula: C₅H₄N₂O₂
MW: 124.1 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: ca 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1
Merck Index: 12,08143
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty