Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-3,4-dichloroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,4-Dichlorophenacyl bromide 2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one 2-Bromo-3',4'-dichloroacetophenone α-Bromo-3,4-dichloroacetophenone

A13365-22 A13365-14 A13365-06 A13365-06 A13365-14 A13365-22
AAA13365-06EA 59.39 USD InStock
AAA13365-06 AAA13365-14 AAA13365-22 CAAAA13365-06 CAAAA13365-22 CAAAA13365-14
ω-Bromo-3,4-dichloroacetophenone 98%
ω-Bromo-3,4-dichloroacetophenone


Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Melting Pt: 55…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051581
CAS Number: 2632-10-2
EINECS: 000-000-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty