Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromocumene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-4-isopropylbenzene4-Bromoisopropylbenzene

A13370-18 A13370-09 A13370-09 A13370-06 A13370-06 A13370-18
AAAA13370-09EA 142.99 USD InStock
AAAA13370-09 AAAA13370-18 CAAAAA13370-09 CAAAA13370-06 CAAAAA13370-18 AAA13370-06
4-Bromocumene 97%
4-Bromocumene


Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 219 °C
Melting Pt: –12 °C
Density: 1.315
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039159
CAS Number: 586-61-8
EINECS: 209-577-5
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty