Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-methyl-2-butene 90+% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Bromo-2-methyl-2-butene3,3-Dimethylallyl bromide

Stabilization: Stabilised with silver

A13388-09 A13388-30 A13388-18 A13388-30 A13388-18 A13388-09
AAA13388-18EA 172.99 USD InStock
AAA13388-18 CAAAA13388-09 AAA13388-09 CAAAA13388-30 CAAAA13388-18 AAA13388-30
1-Bromo-3-methyl-2-butene 90+% stabilized
1-Bromo-3-methyl-2-butene


Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 135 °C
Density: 1.281
Flash Pt: 32 °C(90 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000242
CAS Number: 870-63-3
EINECS: 212-799-5
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty