Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-4-Fluoromandelic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-4-Fluorophenylglycolic acid

A13391-06 A13391-03 A13391-14 A13391-06 A13391-03 A13391-14
AAA13391-03EA 45.39 USD InStock
AAA13391-03 AAA13391-14 CAAAA13391-06 AAA13391-06 CAAAA13391-03 CAAAA13391-14
(±)-4-Fluoromandelic acid 98%
(±)-4-Fluoromandelic acid


Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00044402
CAS Number: 395-33-5
EINECS: 206-899-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty