Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dimethoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3,4-Dimethoxyphenyl)methanol Veratryl alcohol

A13396-36 A13396-22 A13396-22 A13396-14 A13396-36 A13396-14
AAA13396-36EA 457.99 USD InStock
AAA13396-36 AAA13396-14 CAAAA13396-36 AAA13396-22 CAAAA13396-14 CAAAA13396-22
3,4-Dimethoxybenzyl alcohol 97%
3,4-Dimethoxybenzyl alcohol


Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 297…298 °C
Melting Pt: ca 22 °C
Density: 1,176
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty