Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorocinnamic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: (2E)-3-(3-Chlorophenyl)acrylic acid3-(3-Chlorophenyl)acrylic acid

A13416-09 A13416-18 A13416-09 A13416-18 A13416-30 A13416-30
AAA13416-30EA 542.99 USD InStock
AAA13416-30 AAA13416-09 AAA13416-18 CAAAA13416-30 CAAAA13416-18 CAAAA13416-09
3-Chlorocinnamic acid predominantly trans 98+%
3-Chlorocinnamic acid


Formula: C₉H₇ClO₂
MW: 182.61 g/mol
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00004384
CAS Number: 1866-38-2
EINECS: 217-478-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty