Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-3-Hexenoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

A13428-09 A13428-30 A13428-18 A13428-18 A13428-09
AAA13428-18EA 150.99 USD InStock
CAAAA13428-18 AAA13428-18 CAAAA13428-09 AAA13428-09 CAAAA13428-30
trans-3-Hexenoic acid 97%
trans-3-Hexenoic acid


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 206 °C
Melting Pt: 11…12 °C
Density: 0.961
Flash Pt: 99 °C(210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002786
CAS Number: 1577-18-0
EINECS: 216-417-8
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty