Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenyl-1-butanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Phenylbutan-1-ol

A13433-30 A13433-30 A13433-18 A13433-18 A13433-09 A13433-09
AAA13433-18EA 328.99 USD InStock
CAAAA13433-09 AAA13433-18 CAAAA13433-18 AAA13433-09 AAA13433-30 CAAAA13433-30
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol


Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 138…140 °C/14 mm
Density: 0.975
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6
EINECS: 222-128-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty