Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-o-xylene 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methylbenzyl chloride

A13448-36 A13448-22 A13448-36 A13448-22
AAA13448-22EA 82.89 USD InStock
AAA13448-22 AAA13448-36 CAAAA13448-36 CAAAA13448-22
α-Chloro-o-xylene 98+%
α-Chloro-o-xylene


Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 197…199 °C
Melting Pt: 2 °C
Density: 1.081
Flash Pt: 73 °C(163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: "8",III
Merck Index: 14,10091
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty