Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-4-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A13459-14 A13459-14 A13459-36 A13459-36 A13459-22 A13459-22
AAA13459-22EA 135.99 USD InStock
CAAAA13459-14 CAAAA13459-36 AAA13459-22 AAA13459-14 AAA13459-36 CAAAA13459-22
1-Chloro-4-iodobenzene 99%
1-Chloro-4-iodobenzene


Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 226…227 °C
Melting Pt: 52…56 °C
Density: 1,886 g/cm³ (57 °C)
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty