Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Anisic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methoxybenzoic acid

A13513-22 A13513-36 A13513-22 A13513-36
AAA13513-22EA 49.39 USD InStock
AAA13513-22 CAAAA13513-36 CAAAA13513-22 AAA13513-36
o-Anisic acid 98+%
o-Anisic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 99…103 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS Number: 579-75-9
EINECS: 209-447-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty