Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-2-pyridone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pyridine-2,4-diol

A13566-06 A13566-03 A13566-14 A13566-14 A13566-06 A13566-03
AAA13566-06EA 203.99 USD InStock
AAA13566-06 AAAA13566-14 CAAAA13566-06 CAAAAA13566-14 CAAAA13566-03 AAA13566-03
4-Hydroxy-2-pyridone 97%
4-Hydroxy-2-pyridone


Formula: C₅H₅NO₂
MW: 111.1 g/mol
Melting Pt: ca 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006273
CAS Number: 626-03-9
EINECS: 210-924-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty