Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Methylthiazol-5-yl)ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-(2-Hydroxyethyl)-4-methylthiazole 4-Methyl-5-thiazoleethanol 4-Methyl-5-(2-hydroxyethyl)thiazole

A13567-14 A13567-22 A13567-22 A13567-14
AAA13567-22EA 91.79 USD InStock
AAA13567-22 CAAAA13567-14 CAAAA13567-22 AAA13567-14
2-(4-Methylthiazol-5-yl)ethanol 98%
2-(4-Methylthiazol-5-yl)ethanol


Formula: C₆H₉NOS
MW: 143.21 g/mol
Boiling Pt: 135…137 °C/7 mm
Density: 1,196
Flash Pt: 142 °C(287 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005339
CAS Number: 137-00-8
EINECS: 205-272-6
Merck Index: 12,06208
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty