Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Rutin trihydrate 95%

Supplier: Thermo Scientific Chemicals

A13570-14 A13570-14 A13570-22 A13570-36 A13570-36 A13570-22
AAA13570-14EA 42.39 USD InStock
CAAAA13570-14 CAAAA13570-36 CAAAA13570-22 AAA13570-14 AAA13570-36 AAA13570-22
(+)-Rutin trihydrate 95%
(+)-Rutin trihydrate


Formula: C₂₇H₃₀O₁₆.3H₂O
Melting Pt: 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149490
CAS Number: 250249-75-3
EINECS: 205-814-1
Order Now
Specification Test Results
Optical rotation +9° (c=0.5 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty