Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(3-Phenylpropyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

A13590-0B A13590-30 A13590-30 A13590-0B A13590-18 A13590-18
AAA13590-18EA 25.39 USD InStock
AAA13590-18 CAAAA13590-18 CAAAA13590-0B AAA13590-30 CAAAA13590-30 AAA13590-0B
4-(3-Phenylpropyl)pyridine 98%
4-(3-Phenylpropyl)pyridine


Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Boiling Pt: 320…322 °C
Density: 0,853
Flash Pt: 140 °C(284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00047458
CAS Number: 2057-49-0
EINECS: 218-159-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty