Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzoylacetonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(2-Chlorophenyl)-3-oxopropanenitrile

A13611-22 A13611-14 A13611-22 A13611-06 A13611-14 A13611-06
AAA13611-14EA 269.99 USD InStock
AAA13611-14 AAAA13611-22 CAAAA13611-06 CAAAA13611-14 AAA13611-06 CAAAAA13611-22
2-Chlorobenzoylacetonitrile 95%
2-Chlorobenzoylacetonitrile


Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00051624
CAS Number: 40018-25-5
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty