Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Phenylalanine ethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-Phe-OEt·HCl

A13618-22 A13618-14 A13618-06 A13618-22 A13618-06 A13618-14
AAA13618-22EA 296.99 USD InStock
AAA13618-22 CAAAA13618-22 AAA13618-14 AAA13618-06 CAAAA13618-06 CAAAA13618-14
L-Phenylalanine ethyl ester hydrochloride 98%
L-Phenylalanine ethyl ester hydrochloride


Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00012507
CAS Number: 3182-93-2
EINECS: 221-673-9
Order Now
Specification Test Results
Optical rotation +36° (c=5 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty