Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Anisohydrazide 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Methoxybenzene-1-carbohydrazide 2-Methoxybenzhydrazide 2-Methoxybenzohydrazide 2-Methoxybenzoic acid hydrazide 2-Methoxybenzoic hydrazide 2-Anisohydrazide

A13619-18 A13619-09 A13619-30 A13619-18 A13619-09 A13619-30
AAA13619-18EA 513.99 USD InStock
CAAAA13619-30 AAA13619-18 CAAAA13619-09 AAA13619-09 CAAAA13619-18 AAA13619-30
o-Anisohydrazide 98+%
o-Anisohydrazide


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00014755
CAS Number: 7466-54-8
EINECS: 231-260-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty