Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: m-Hydroxybenzoic Acid

A13628-36 A13628-22 A13628-36 A13628-22
AAA13628-36EA 204.99 USD InStock
CAAAA13628-22 AAA13628-36 AAA13628-22 CAAAA13628-36
3-Hydroxybenzoic acid 99%
3-Hydroxybenzoic acid


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 201…203 °C
Density: 1,473 g/cm³ (20 °C)
Flash Pt: 220 °C(428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty